Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tube Jockey

Supplier: MARKETLAB, INC.

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

1000 4000 5000
470236-380EA 69.49 USD InStock
470236-380 470236-376 470236-378
Tube Jockey
Clinical Diagnostic Supplies Clinical Instruments
The Tube Jockey keeps tubes within reach.

  • Always have a safe place to put tubes
  • Tube holder securely attaches to wristband with Velcro®
  • Holds both 13 mm and 16 mm tubes
  • Buy as a kit or as individual parts

No more twisting and reaching for tubes. With the Tube Jockey, users are free to draw the patient in the safest, most convenient, and most organized position possible. Now, no matter where you go to draw, you will always have a safe place to put your tubes and they will always be in full view.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty