Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Ward's® Magnetic Hotplate Stirrer
Supplier: VWR International
Ward's®
8050221010
470236-626EA 399 USD InStock
470236-626
Ward's® Magnetic Hotplate Stirrer
Stirrers Magnetic Hotplate Stirrers
This cost-efficient, durable hotplate stirrer meets or exceeds all leading safety standards.
  • Temperature to 340 °C
  • Stirring speed up to 1500 rpm
Safety circuits provide overheating protection.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty