Would you like to log in again?
Raspberry Pi 3 Model B Project Board
Supplier: Rasberry Pi
31AC4864 77Y6520
470237-718EA 69.69 USD InStock
470237-718 470237-716
Raspberry Pi 3 Model B Project Board
Educational Materials Physics Educational Materials Applied Physics Learning Activities
Pocket Sized, Fun and Easy to Use.
  • Free online training available
  • Access to the Raspberry Pi community of educators
  • 100+ projects available
  • Quad Core 1.2 GHz Broadcom BCM2837 64 bit CPU
  • 1GB RAM
  • BCM43438 WiFi and Bluetooth Low Energy (BLE) on board
  • 40-pin Extended GPIO
  • 4× USB 2 ports
  • 4 Pole stereo output and composite video port
  • Full size HDMI
  • CSI camera port for connecting a Raspberry Pi camera
  • DSI display port for connecting a Raspberry Pi touchscreen display
  • Micro SD port for loading your operating system and storing data
The Raspberry Pi 3 Model B builds upon the features of its predecessors with a new, faster processor on board to increase its speed. It also features WiFi and Bluetooth Low Energy capabilities to enhance the functionality and the ability to power more powerful devices over the USB ports.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty