Would you like to log in again?
Exploring Meteorology
Supplier: ALDON CORP SE
IS8706
470237-788KT 67 USD InStock
470237-788
Exploring Meteorology
Educational Materials Earth Science Educational Materials Meteorology Learning Activities
Learn how to meteorologists predict the weather.
  • Perfect for middle and high school students
  • Aligned to NGSS ESS2.C and ESS2.D
Weather impacts everyone, everyday in some way and the meteorologist that study weather patterns are our best source for information. This experiment provides students with the experience of being a meteorologist. It includes studies of pressure changes and creating their own barometer. Studies of wind speed and creating their own anemometer and, the skills to confidently read a weather map. *Bonus* this experiment includes a demonstration of how clouds form by allowing the instructor to form a vapor cloud. Experiment includes enough material for 15 groups of 2 to perform the tasks. The demonstration portion has enough material to perform the demonstration 5 times.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty