Would you like to log in again?
Forensic Drug Testing: A Simulated Immunoassay
Supplier: ALDON CORP SE
IS9023
470237-798KT 81.3 USD InStock
470237-798
Forensic Drug Testing: A Simulated Immunoassay
Educational Materials Forensics Educational Materials Trace Evidence Learning Activities
Real-Life Forensic Urine Analysis.
  • Perfect for middle school students
  • Aligned to NGSS LS1.A, PS1.A and PS1. B
Drug testing comes in two forms, presumptive and confirmatory. Because of the cost associated with confirmatory testing, a presumptive test is often employed first. While there are various forms of presumptive drug testing, two of the most common techniques are immunoassay testing and thin-layer chromatography. In this activity, students will perform a simulated immunoassay test on several urine samples for the detection of the cocaine metabolite benzoylecgonine. All urine samples and testing reagents are simulated to avoid the potential hazards of actual biological specimens while still providing realistic results.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty