Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium Chloride

Supplier: ALDON CORP SE
AA0243-500ML AA0235-500G AA0241-500ML AA0235-100GR AA0235-500GR AA0235-2.5KG
470225-754EA 9.75 USD InStock
470225-754 470300-196 470300-194 470300-192 470300-190 470300-188
Ammonium Chloride
Chemicals for Science Education
Formula: NH4Cl
Density: 1.527 g/mL
Boiling Point: Sublimes at 340°C
Freezing Point: 520°C
Solubility: Water and Glycerol
Synonyms: Ammonium Muriate, Sal Ammoniac
Shelf Life: 12 Months

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Caution: This product is an irritant.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty