Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Ascorbic Acid

Supplier: ALDON CORP SE
AA0425-SCAP AA0425-CAPS AA0425-100G
470300-284EA 12.45 USD InStock
470300-284 470300-282 470300-286
Ascorbic Acid
Chemicals for Science Education
CAS Number: 50-81-7
Formula: C6H8O6
Density: 1.65 g/mL
Boiling and Freezing Point: Decomposes, 190-192°C
Synonyms: Vitamin C
Shelf Life: 36 Months

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty