Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Barium Nitrate

Supplier: ALDON CORP SE

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

BB0055-CAP BB0055-CAPS BB0055-100G BB0055-500G BB0061-500ML BB0055-500GR BB0055-100GR
470300-338EA 56.69 USD InStock
470300-338 470300-348 470300-336 470300-346 470300-344 470300-342 470300-340
Barium Nitrate
Chemicals for Science Education
CAS Number: 10022-31-8
Formula Weight: 261.35
Formula: Ba(NO3)2
Hazard Info: Oxidizer, Toxic
Density (g/mL): 3.24
Boiling Point (°C): Decomposes
Freezing Point (°C): 592
Solubility: Water
Synonyms: Barium Dinitrate, Nitric Acid Barium Salt
Shelf Life (months): 36
Storage: Yellow

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty