Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Buffer Solution

Supplier: ALDON CORP SE

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

BB0409-500ML BB0404-1L BBB410-500ML_B BB0405-500ML BB0407-1L BB0403-1L BB0403-500ML BB0409-1L BB0406-500ML BB0408-1L BB0402-1L BB0410-500ML BB0410-1L BBR404-500ML BB0412-500ML BB0405-1L BB0408-500ML BBY407-500ML BB0411-500ML BB0406-1L
470300-504EA 15.45 USD InStock
470300-504 470300-502 470300-482 470300-460 470300-480 470300-490 470300-468 470300-500 470300-466 470300-488 470300-476 470300-498 470300-464 470300-486 470300-474 470300-496 470300-462 470300-484 470300-472 470300-494
Buffer Solution
Buffers Cell Culture Buffers
Buffer solution has a shelf life of 24 months

  • Density: Approximately 1 g/mL
  • Boiling Point: 100°C
  • Freezing Point: 0
  • Shelf Life: 24 Months
  • Storage: Green

Its density is 1 g/mL.

Buffer solution has a boiling point of 100°C and a freezing point of 0°C.

Caution: Caution:This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty