Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl Alcohol Denatured

Supplier: ALDON CORP SE
EE0076-CS/2 EE0076-500ML EE0076-3.8LR EE0076-PK/2 EE0076-500MLR EE0075-1LR EE0076-1L EE0075-500MLR EE0069-3.8L EE0075-3.8SCR EE0075-3.8LR EE0076-20L EE0076-3.8L EE0076-3.8SC EE0075-20LR EE0076-20L EE0076-3.8SCR
470301-072EA 45.25 USD InStock
470301-072 470301-050 470301-060 470301-052 470301-074 470108-538 470301-062 470301-070 470301-058 470301-068 470301-046 470301-054 470301-064 470301-056 470301-044 470301-066 470233-566
Ethyl Alcohol Denatured
Chemicals for Science Education
CAS Number: Mixture
Formula: Mixture
Density: 0.794 g/mL
Boiling and Freezing Point: 78°C, -114°C
Solubility: Miscible with Water and Organic Solvents
Synonyms: Denatured Alcohol, Ethyl Alcohol Anhydrous, Ethanol
Shelf Life: 36 Months

  • Aligned with GLOBE protocols

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Caution: This product is flammable and toxic.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty