Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric Acid

Supplier: ALDON CORP SE
HH0100-1L HH0120-1L HH0158-3ML HH0160-500ML HH0093-500ML HH0099-1L HH0090-PK2 AD-HH0171P-PK/6 HH0094-500ML HH0170-1L HH0071-CONC HH0171-500MLR HH0170-PK/2 HH0171-PK/4R HH0090-1L HH0090-3.8L HH0170-500ML HH0171-100MLR HH0168-3.8L AD-HH0171P-2.5L HH0093-1L HH0071-2.5L HH0100-3.8L HH0171-PK2R HH0165-500ML HH0090-500ML HH0171-PK/2R HH0171-2.5LR R2820F-AMP HH0168-1L HH0095-500ML
470301-260EA 10.1 USD InStock
470301-260 470301-222 470301-266 470301-244 470301-224 470301-246 470301-268 470301-262 470301-240 470301-242 470301-264 470354-380 470229-790 470301-248 470301-226 470301-228 470301-250 470113-126 470301-234 470301-256 470301-236 470301-258 470301-230 470301-252 470301-254 470301-232 470229-788 470225-696 470301-238 470301-218
Hydrochloric Acid
Chemicals for Science Education
CAS Number: 7647-01-0
Formula: HCl
Density: 1.16 mL
Boiling and Freezing Point: 81.11 to 85°C, -45°C
Solubility: Water
Synonyms: Muriatic Acid, Hydrogen Chloride
Shelf Life: 24 Months

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Caution: This product is corrosive and toxic.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty