Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Iron(III) Chloride

Supplier: ALDON CORP SE
FF0100-500GR FF0100-100GR FF0101-500ML FF0102-CONC FF0102-100ML FF0100-500G FF0100-100G
470301-382EA 18.15 USD InStock
470301-382 470301-392 470301-388 470301-394 470301-386 470301-584 470301-396
Iron(III) Chloride
Chemicals for Science Education
CAS Number: 7705-08-0
Formula Weight: 162.21
Formula: FeCl3
Hazard Info: Irritant, Corrosive, Hygroscopic
Density (g/mL): 2.80-2.90
Boiling Point (°C): 307
Freezing Point (°C): 307
Solubility: Water
Synonyms: Ferric Chloride Anhydrous
Shelf Life (months): 12
Storage: White

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty