Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Magnesium Oxide

Supplier: ALDON CORP SE
MM0110-500G MM0110-100G MM0120-500G
470301-678EA 56.59 USD InStock
470301-678 470301-674 470301-676
Magnesium Oxide
Chemicals for Science Education
CAS Number: 1309-48-4
Formula Weight: 40.30
Formula: MgO
Hazard Info: Toxic
Density (g/mL): 3.65-3.75
Boiling Point (°C): 3600
Freezing Point (°C): 2800
Solubility: Acids and Slightly in Water
Synonyms: Magnesia, Calcinated Magnesia, Calcined Brucite
Shelf Life (months): 12
Storage: Green

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty