Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nickel Nitrate 6-Hydrate

Supplier: ALDON CORP SE
NN0159-CONC NN0150-500G NN0161-500ML NN0150-100G
470301-858EA 13.95 USD InStock
470301-858 470301-854 470301-856 470301-860
Nickel Nitrate 6-Hydrate
Chemicals for Science Education
CAS Number: 13478-00-7
Formula Weight: 290.81
Formula: Ni(NO3)2·6H2O
Hazard Info: Suspected Human Carcinogen, Toxic
Density (g/mL): 2.065
Boiling Point (°C): 138
Freezing Point (°C): 57
Solubility: Water, Alcohol, and Ammonium Hydroxide
Synonyms: Nickel Nitrate Hexahydrate, Nickelous Nitrate 6-Hydrate
Shelf Life (months): 12
Storage: Yellow

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty