Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium Chromate

Supplier: ALDON CORP SE
PP0480-100GR PP0495-500ML PP0480-100G PP0480-500G PP0480-500GR PP0494-50ML
470302-094EA 11.5 USD InStock
470302-094 470225-704 470302-098 470302-096 470302-102 470302-100
Potassium Chromate
Chemicals for Science Education
CAS Number: 7789-00-6
Formula Weight: 194.21
Formula: K2CrO4
Hazard Info: Toxic
Density (g/mL): 2.732
Freezing Point (°C): 975
Solubility: Water
Synonyms: Dipotassium Chromate, Neutral Potassium chromate
Shelf Life (months): 36
Storage: Blue

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty