Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium Permanganate

Supplier: ALDON CORP SE
PP0700-500G PP0700-100GR PP0711-500ML PP0700-PK/4 PP0711-1L PP0700-100G PP0700-500GR
470302-228PK 23.25 USD InStock
470302-228 470302-216 470302-238 470302-214 470302-236 470302-220 470302-212
Potassium Permanganate
Chemicals for Science Education
CAS Number: 7722-64-7
Formula Weight: 158.04
Formula: KMnO4
Hazard Info: Oxidizer, Explosive, Irritant
Density (g/mL): 2.7032
Boiling Point (°C): Decomposes
Freezing Point (°C): Decomposes
Solubility: Water
Synonyms: Chameleon Mineral
Shelf Life (months): 36
Storage: Yellow

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty