Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium Phosphate Monobasic

Supplier: ALDON CORP SE
PP0730-100GR PP0730-500GR PP0730-100G PP0730-500G
470302-248EA 13.45 USD InStock
470302-248 470302-252 470302-250 470302-254
Potassium Phosphate Monobasic
Chemicals for Science Education
CAS Number: 7778-77-0
Formula Weight: 136.09
Formula: KH2PO4
Density (g/mL): 2.338
Freezing Point (°C): 253
Solubility: Water
Synonyms: Potassium Dihydrogen Phosphate
Shelf Life (months): 12
Storage: Green

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty