Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium Thiocyanate

Supplier: ALDON CORP SE
PP0801-500ML PP0802-CONC PP0790-500G PP0790-100G PP0790-500GR
470302-282EA 38.35 USD InStock
470302-282 470302-280 470302-286 470302-278 470302-276
Potassium Thiocyanate
Chemicals for Science Education
CAS Number: 333-20-0
Formula Weight: 97.18
Formula: KSCN
Hazard Info: Toxic
Density (g/mL): 1.886
Boiling Point (°C): 500
Freezing Point (°C): 173
Solubility: Water, Alcohol, and Acetone
Synonyms: Thiocyanic Acid Potassium Salt, Potassium Sulfocyanate, Potassium Rhodanide
Shelf Life (months): 12
Storage: Green

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty