Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium Acetate Anhydrous

Supplier: ALDON CORP SE
SS0230-500G SS0229-500ML SS0230-2.5KG SS0230-100G SS0230-500GR SS0228-500ML
470302-424EA 38 USD InStock
470302-424 470045-542 470302-422 470225-794 470302-420 470225-674
Sodium Acetate Anhydrous
Chemicals for Science Education
CAS Number: 127-09-3
Formula: NaC2H3O2
Density: 1.53 g/mL
Freezing Point: 324°C
Solubility: Water and Alcohol
Synonyms: Acetic Acid Sodium Salt
Shelf Life: 12 Months

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Caution: This product is toxic.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty