Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium Borate, 10-Hydrate

Supplier: ALDON CORP SE
SS0320-100GR SS0320-100GR SS0320-500G SS0320-500GR SS0320-500G
470302-456EA 8.75 USD InStock
470302-456 470233-596 470233-648 470302-454 470302-452
Sodium Borate, 10-Hydrate
Chemicals for Science Education
CAS Number: 1303-96-4
Formula: Na2B4O7 10H2O
Density: 1.73 g/mL
Freezing Point: 62 °C
Solubility: Water and Glycerin
Synonyms: Sodium Tetraborate, Borax
Shelf Life: 12 Months

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty