Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium Bromide

Supplier: ALDON CORP SE
SS0340-500GR SS0353-500ML SS0340-100G SS0340-500G SS0352-1L SS0351-500ML SS0340-100GR SS0349-500ML
470225-732EA 22.9 USD InStock
470225-732 470302-458 470225-746 470302-462 470302-460 470225-708 470302-466 470302-464
Sodium Bromide
Chemicals for Science Education
CAS Number: 7647-15-6
Formula Weight: 102.90
Formula: NaBr
Hazard Info: Toxic
Density (g/mL): 3.203
Boiling Point (°C): 1391
Freezing Point (°C): 755
Solubility: Water and Alcohol
Synonyms: Sedoneual
Shelf Life (months): 12
Storage: Green

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty