Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium Chloride

Supplier: ALDON CORP SE
SS0430-CAPS SS0441-500ML SS0440-1L SS0430-2.5KG SS0450-2.5KG SS0437-1L SS0430-2.5KGR SS0442-500ML SS0430-500GR SS0430-12KG SS0442-50ML SS0436-500ML SS0450-500G SS0430-735G
470302-502EA 7.1 USD InStock
470302-502 470302-512 470302-522 470302-500 470302-506 470302-516 470302-504 470225-730 470225-796 470225-798 470302-494 470302-510 470302-498 470302-520
Sodium Chloride
Chemicals for Science Education
CAS Number: 7647-14-5
Formula: NaCl
Density: 2.16 g/mL
Boiling and Freezing Point: 1465°C, 801°C
Shelf Life: 36 Months

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty