Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium Chromate Anhydrous

Supplier: ALDON CORP SE
SS0470-500G SS0470-100G SS0471-500ML
470302-524EA 19 USD InStock
470302-524 470225-698 470302-526
Sodium Chromate Anhydrous
Chemicals for Science Education
CAS Number: 7775-11-3
Formula Weight: 161.99
Formula: Na2CrO4
Hazard Info: Toxic
Density (g/mL): 2.723
Freezing Point (°C): 792
Solubility: Water and Alcohol
Synonyms: Disodium Chromate(VI), Chromate of Soda
Shelf Life (months): 36
Storage: Blue

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty