Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium Dodecyl Sulfate

Supplier: ALDON CORP SE
SS0650-500G SS0647-100ML SS0650-25GR SS0648-50ML SS0650-100G
470302-612EA 9.6 USD InStock
470302-612 470302-610 470302-616 470302-614 470302-618
Sodium Dodecyl Sulfate
Chemicals for Science Education
CAS Number: 151-21-3
Formula: CH3CH211OSO3Na
Freezing Point: 204 to 207°C
Solubility: Water and Alcohol
Synonyms: Sodium Lauryl Sulfate, SDS
Shelf Life: 36 Months

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty