Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium Sulfate

Supplier: ALDON CORP SE
SS0830-2.5KG SS0830-500GRG SS0841-50ML SS0830-500G SS0830-500GRC SS0841-500ML SS0830-500GP SS0828-500ML SS0842-500ML
470302-700EA 9.75 USD InStock
470302-700 470302-688 470302-704 470302-702 470302-692 470302-690 470302-696 470225-672 470302-694
Sodium Sulfate
Chemicals for Science Education
CAS Number: 7757-82-6
Formula Weight: 142.04
Formula: Na2SO4
Density (g/mL): 2.698
Boiling Point (°C): >1000
Freezing Point (°C): 888
Solubility: Water
Synonyms: Disodium Sulfate
Shelf Life (months): 36
Storage: Green

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty