Would you like to log in again?
Stain Set, Basic
Supplier: ALDON CORP SE
SS0930-KIT
470302-740EA 69.45 USD InStock
470302-740
Stain Set, Basic
Chemicals for Science Education
Includes: Bismarck Brown, Carmine Alum (Alcoholic), Crystal Violet, Eosin (Yellowish), Hematoxylin (Alcoholic), Malachite Green, Methyl Green, Methylene Blue, Neutral Red, Safranin
Shelf Life (months): 36
Storage: Red
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty