Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sudan III

Supplier: ALDON CORP SE
SS1040-10G SS1040-10G SS1040-25G SS1050-100ML
470302-822EA 25.4 USD InStock
470302-822 470302-820 470109-390 470302-824
Sudan III
Chemicals for Science Education
CAS Number: 85-86-9
Formula: C22H16N4O
Density: 0.34 g/mL
Freezing Point: 100 °C
Solubility: Alcohol
Synonyms: Solvent Red 23 C.I. No. 26100
Shelf Life: 36 Months

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Caution: This product is flammable.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty