Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sulfuric Acid

Supplier: ALDON CORP SE
SS1115-2.5L SS1128-500ML SS1117-500ML SS1125-1L SS1126-500ML SS1122-1L SS1127-1L SS1118-500ML SS1126-1L AD-SS1118P-PK/6 AD-SS1118P-2.5L SS1121-1L SS1120-500ML SS1118-2.5LR SS1118-CONC SS1116-1L SS1118B-PK/4R SS1127-1L SS1125-500ML
470302-866EA 24 USD InStock
470302-866 470302-854 470302-876 470302-864 470302-852 470302-858 470302-868 470302-878 470302-856 470225-682 470045-604 470354-382 470109-408 470354-388 470302-862 470302-872 470302-850 470302-860 470302-870
Sulfuric Acid
Chemicals for Science Education
CAS Number: 7664-93-9
Formula: H2SO4
Density: 1.84 g/mL
Boiling and Freezing Point: 275-325°C, 11°C
Solubility: Water
Synonyms: Hydrogen Sulfate, Battery Acid
Shelf Life: 12 Months

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Caution: This product is corrosive.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty