Would you like to log in again?
Ward's® AP Chemistry Investigation 14: Acid-Base Titrations: How does Chemical Structure Affect pH?
Supplier: ALDON CORP SE
470303-022
470303-022KT 48.59 USD InStock
470303-022
Ward's® AP Chemistry Investigation 14: Acid-Base Titrations: How does Chemical Structure Affect pH?
Educational Materials Chemistry Educational Materials Applied Chemistry Learning Activities
Meets AP Science Practices, 1, 2, 3, 4, 5, 6, and 7
  • Addresses Big Ideas 1 & 6
  • Materials provided for 30 students working in pairs
  • Investigation includes updated literature aligned with new AP Chemistry curriculum standards
In this lab, acid-base titrations will be performed in order to investigate the relationship between pH, concentration, and the structure of acids and bases. Students will carry out multiple acid-base titrations and will use a pH meter to monitor the pH of the resulting solutions. Students will then construct titration curves. Using known information and the data that was gathered, students will ultimately be able to determine the concentration of the acid or base, the ionization constant, and the percent ionization. Includes hydrochloric acid, acetic acid, sodium hydroxide, and phenolphthalein. Requires pH meter, burette, ring stand/clamps, magnetic stirrer, and magnetic stir bar, available separately. Ward's® DataHub is not required to complete activity.
AP and Advanced Placement Programs are registered trademarks of the College Entrance Examination Board, which was not involved in the production of and does not endorse these products.
Ordering information: Activity includes Hydrochloric Acid, Acetic Acid, Sodium Hydroxide, and Phenolphthalein. Materials required but not provided are Graduated Cylinders, Erlenmeyer Flasks or Beakers, Vials/Test Tubes, Pipettes, Burettes, Ring Stands/Clamps, pH Meters, DI Water, Gloves, Safety Goggles, and Lab Aprons.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty