Would you like to log in again?
Ward's® AP Chemistry Investigation 15: Buffering: Will that Product Act as a Buffer?
Supplier: ALDON CORP SE
470303-024
470303-024KT 82.29 USD InStock
470303-024
Ward's® AP Chemistry Investigation 15: Buffering: Will that Product Act as a Buffer?
Educational Materials Chemistry Educational Materials Applied Chemistry Learning Activities
Meets AP Science Practices 4, 5, and 6
  • Addresses Big Ideas 1 & 6
  • Materials provided for 30 students working in pairs
  • Investigation includes updated literature aligned with new AP Chemistry curriculum standards
In this lab, students will observe the buffering abilities of a number of different household products. Students will use the concepts of acid-base chemistry, titrations, pH, and buffers in order to determine if certain household products will act as buffers, and if so which chemicals are present. Titration curves will be constructed for known chemicals and then compared to the titration curves produced for the household products. Includes sodium hydroxide, citric acid, diet coke, diet lemon-lime soda, lemonade, grape juice, phosphoric acid, and acetic acid. Requires pH meter, ring stand/clamps, magnetic stirrer, magnetic stir rod, and burette, available separately. Ward's® DataHub not required to complete activity.
AP and Advanced Placement Programs are registered trademarks of the College Entrance Examination Board, which was not involved in the production of and does not endorse these products.
Includes:
Sodium Hydroxide
Hydrochloric Acid
Citric Acid
Phosphoric Acid
Acetic Acid
Diet Coke
Diet Sprit
Lemonade
Grape Juice
Required but Not Provided: pH Meter, Beakers, Pipettes, Graduated Cylinders, Erlenmeyer Flasks, Ring Stands/Clamps, Burettes, DI Water, Gloves, Safety Goggles, Lab Aprons.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty