Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zinc Chloride

Supplier: ALDON CORP SE
ZZ0069-CONC ZZ0060-100G ZZ0070-500ML ZZ0060-500G
470303-080EA 31.7 USD InStock
470303-080 470303-074 470303-076 470303-078
Zinc Chloride
Chemicals for Science Education
CAS Number: 7646-85-7
Formula: ZnCl2
Density: 2.91 g/mL
Boiling Point: 732°C
Freezing Point: 290°C
Solubility: Water and Alcohol
Shelf Life: 36 Months

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Caution: This product is corrosive.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty