Would you like to log in again?
König® Green Band Fiber Optic Laryngoscopes Blades
Supplier: MEDLINE INDUSTRIES, INC.
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
MDS0425125 MDS0425115 MDS0423520 MDS0423503 MDS0425120 MDS0423510 MDS0425110 MDS0423505 MDS0423515 MDS0423525
470303-324EA 117.55 USD InStock
470303-324 470303-312 470303-326 470303-314 470303-328 470303-316 470303-318 470303-320 470303-322 470303-310
König® Green Band Fiber Optic Laryngoscopes Blades
Razors and Blades
The Miller and Macintosh blades are compatible with any of the green band handles. Each blade is made from surgical grade stainless steel and has a large enclosed Fiberoptic core. The blades contain no crevices to trap bioburden and are easily decontaminated.
  • Blades have a large glass fiber bundle for maximum light
  • Light in handle keeps the blades from heating up
These LED Laryngoscopes keep you prepared for any situation. With three available handles to fit your need. Each handle has a knurled texture for a secure grip, even in the most difficult situations, and utilizes a metal collar to direct the maximum amount of light from the LED into the Fiberoptic bundle. The bright white LED lighting will not dim as your batteries run down but will stay bright right up until the end.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty