Would you like to log in again?
STEM Engineering: Biomaterials to Make Your Own Contact Lenses
Supplier: ALDON CORP SE
IS3402
470303-576KT 97.65 USD InStock
470303-576
STEM Engineering: Biomaterials to Make Your Own Contact Lenses
Educational Materials Chemistry Educational Materials Consumer Chemistry Learning Activities
Biomedical Engineering and Design.
  • Real life biomedical situation
  • Enough materials for 15 groups of students
Engineers play an integral role in the process of finding a "perfect" material or ratio of materials to maximize the desired properties and decrease the amount of negative effects of other properties. For example, when developing the perfect material for use in contacts, an engineer will have to test many different materials before finding the right one. Therefore, testing a hydrogel, which is favorable for all three of the main properties needed for a contact lens to function properly seems to be logical. A hydrogel is a polymer similar to plastic, which has favorable optical properties and favorable flexibility/strength but like paper and glasses/ceramics, it is hydrophilic, which is necessary for a contact lens to function properly.
The main job of a biomedical engineer developing contact lenses today is to make a hydrogel and find the perfect "ratio" of polymer to water so that the composition of the hydrogel is hydrophilic enough to maintain a wet environment in the eye but not too hydrophilic so that the contact begins to swell and change size and shape. Students will engineer using 2 different materials, the correct material and concentration that has similar properties as a contact lens with discovering the correct optical properties, tensile strength and hydrophobicity.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty