Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ecoline Series Microscopes, National Optical & Scientific SE

Supplier: National Optical & Scientific Instruments
National®
D-ELM D-ELDB D-ELB
470303-742EA 530.25 USD InStock
470303-742 470303-744 470303-746
Ecoline Series Microscopes, National Optical & Scientific SE
Microscopes
These Ecoline Series Microscopes deliver excellent clarity with standard achromatic lenses and normal flatness of field.

  • Widefield eyepieces
  • Quad objective turret
  • 3.4 V 0.06 watt LED
  • 4.5 V DC auto switch power adapter

DIN standard 4×(0.10 N.A.), 10×(0.25 N.A.), 40×R(0.65 N.A.) and 100×R oil immersion (1.25 N.A.) color-coded objectives.

Focusing: Coaxial coarse and fine focusing controls, coarse has tension adjustment, fine is graduated.

Low profile, ball-bearing mounted mechanical specimen holder with low-position coaxial controls.

Frame: Rugged cast aluminum, gray enamel finish; Base dimensions: 140×210 mm (51/2×81/4)"; Height: 350 mm (133/4)".

Packaging: Molded styrofoam container with vinyl dustcover.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty