Would you like to log in again?
Go Direct™ CO₂ Gas Sensor
Supplier: VERNIER SOFTWARE & TECH SE
GDX-CO2
470303-908EA 224.99 USD InStock
470303-908
Go Direct™ CO₂ Gas Sensor
Data Loggers
The Go Direct CO₂ Gas Sensor measures gaseous carbon dioxide concentration levels, air temperature, and relative humidity.
  • Measure the change in gas concentration during cellular respiration
  • Measure the change in gas concentration during photosynthesis
  • Study the effect of temperature on cell respiration or metabolism of organisms
  • Investigate artificial selection in yeast by comparing respiration rates of different strains
Monitor changes in carbon dioxide, temperature, and relative humidity easily with the Go Direct CO2 Gas Sensor. This sensor includes built-in temperature compensation and humidity protection. A 250 ml Nalgene bottle is included for running controlled experiments with small plants and animals.
The Go Direct robust rechargeable battery offers long battery life and provides always-ready operation. Monitor battery life directly from our free Graphical Analysis™ 4 app. Should the battery run low, simply connect the sensor to the charging cable and keep on collecting-no interruptions or inconvenience to you or your students.
Our Go Direct sensors connect directly to your mobile device, Chromebook™, or computer using our free Graphical Analysis™ 4 app-no additional equipment or software purchases are necessary. Go Direct sensors can be used wired via USB or wirelessly via Bluetooth wireless technology, allowing you to choose the solution best for your classroom or laboratory.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty