Would you like to log in again?
Spouting Cylinder
Supplier: United Scientific Supplies
Ward's®
CYSP01
470303-934EA 30.45 USD InStock
470303-934
Spouting Cylinder
Educational Classroom Kits and Activities
Demonstrate Torricelli’s theorem to show the relationship between water pressure and depth.
  • Clear design
  • Three nozzles at different heights
This clear acrylic cylinder includes three vertically aligned spouting nozzles positioned 43/4" apart.
Each spouting nozzle also includes a stopper so pressure at each height can be shown individually.
A filler hose barb at the top can be used to quickly refill the cylinder or maintain a constant water depth.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty