Would you like to log in again?
Density Cube Set of 24
Supplier: United Scientific Supplies
DCSET24
470303-938EA 203.85 USD InStock
470303-938
Density Cube Set of 24
Educational Materials Physics Educational Materials Physics Fundamentals Learning Activities
Observe the density across different materials and sizes.
  • Foam storage space
  • Includes a Vernier caliper
This set includes three different sized cubes of eight materials commonly used in density experiments. Each material is represented by a 0.5, 1 and 1.5" cube. Materials included are acrylic, aluminum, brass, copper, nylon, polypropylene, PVC and steel. Set includes a reference table showing density values of individual specimens.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty