Would you like to log in again?
Pendulum Investigation Kit
Supplier: United Scientific Supplies
Ward's®
PNLKIT
470304-028EA 72.1 USD InStock
470304-028
Pendulum Investigation Kit
Educational Materials Physics Educational Materials Properties of Matter Learning Activities in Physics
Understanding pendulum motion is fundamental to the study of vibrations and waves.
  • Includes three different pendulum balls
  • Uniquely design pendulum clamp holds three at once
This unique kit includes everything necessary for a full exploration of the basic features of pendulum motion in a complete but economical set.
Students can assess the principal factors of mass, length, and amplitude and even venture into a consideration of air resistance.
The individual components are of proven design - the drilled balls have a larger hole on one end than the other to allow supension at the center of mass, and the triple support features clamps that make adjusting the cord length precise and easy.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty