Would you like to log in again?
Rectangular Refraction Cell
Supplier: United Scientific Supplies
Ward's®
RCRC01
470304-030EA 8.95 USD InStock
470304-030
Rectangular Refraction Cell
Educational Materials Physics Educational Materials Optics Learning Activities
High Quality for the Classroom.
  • Clear acrylic
  • Uniform wall thickness
This clear acrylic rectangular refraction cell is excellent for showing refraction and internal reflection in water, and to measure its refractive index. The cell has uniform wall thickness to eliminate distortions.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty