Would you like to log in again?
Radiation Can Set
Supplier: United Scientific Supplies
Ward's®
RDCN02
470304-032KT 21.35 USD InStock
470304-032
Radiation Can Set
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
High Quality for the Classroom.
  • Includes activity guide
Two identical sized screw cap metal cans (one smooth metal and other painted dull black) to demonstrate the difference in cooling rates of hot water inside the cans due to different surfaces.
The difference in the cooling rates is attributed to the thermal radiation emitted by the two cans.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty