Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Sling Psychrometer
Supplier: United Scientific Supplies
Ward's®
SPCRKIT-PK/15 SPCRKIT
470304-042PK 102.6 USD InStock
470304-042 470304-040
Sling Psychrometer
Hygrometers Psychrometers
Use this simple apparatus to measure relative humidity.
  • Requires assembly
The thermometers mount on a wooden handle so they can be spun to obtain temperature readings.
Ordering information: Includes an activity guide with relative humidity calculation table. Includes two plastic-backed Celsius thermometers, one for dry bulb measurements and the other for wet bulb measurements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty