Would you like to log in again?
Buret Rack
Supplier: EISCO SCIENTIFIC LLC
VWR®
BURRCK12
470304-900EA 53.6 USD InStock
470304-900
Buret Rack
Racks Tube Racks
Safely store up to 12 burets.
  • Natural wood
  • Sturdy design
This buret rack accepts 12 burets of 50 mL capacity, up to 0.75" (2 cm) in diamater. The sturdy design allows burets to be fitted in and removed very easily. Optional brackets allow for the rack to be mounted to the wall. Rack dimensions are 32 x 3 x 23.5".
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty