Would you like to log in again?
Quantum Standard Levitators
Supplier: QUANTUM LEVITATION LTD
Ward's®
11-2 73-459 11-3 11-1
470306-192EA 852.25 USD InStock
470306-192 470306-212 470306-188 470306-190
Quantum Standard Levitators
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
The Quantum Levitators are intended for science demonstrations.
  • Easy to use
  • Highly durable
The levitators are provided with 6 months full no-questions-asked warranty. The Quantum Levitators are able to carry only their own weight plus liquid nitrogen.
The Starter Kit is the most fundamental, entry level for classroom demonstration of Quantum Levitation and Flux Pinning. Perfect for high-school students as well as university undergraduates; the kit is easy to use, highly durable and portable. Use the rectangle magnetic setup to witness the Quantum Locking phenomena and the round magnetic setup to demonstrate frictionless motion.
When used with the new resistance thermometer equipped Ed levitator, the handheld magnetic device serves as a unique experimentation platform where quantum phenomena such as the Meissner effect and flux pinning can be investigated.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty