Would you like to log in again?
Quantum Levitation MiniMaglev Kit
Supplier: QUANTUM LEVITATION LTD
Ward's®
304-112 305-112-122
470306-208EA 2564.15 USD InStock
470306-208 470306-206
Quantum Levitation MiniMaglev Kit
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Experience Quantum Levitation with this portable, light weight and easy to use maglev track.
  • Easy to use
  • Highly durable
The track uses super-strong NdFeB magnets in a unique double rings design to achieve maximal levitation height. Demonstrate frictionless motion in both levitation and suspension.
The handheld magnetic device (Starter Kit) can be passed around for a captivating hands-on experience.
Excellent for science demonstrations in schools and universities, undergraduate and high-school experiments in quantum and classical physics (energy conservation, harmonic oscillator).
The Starter Kit is the most fundamental, entry level for classroom demonstration of Quantum Levitation and Flax Pinning. Perfect for high-school students as well as university undergraduates; the kit is easy to use, highly durable and portable. Use the rectangle magnetic setup to witness the Quantum Locking phenomena and the round magnetic setup to demonstrate frictionless motion.
When used with the new resistance thermometer equipped Ed levitator, the handheld magnetic device serves as a unique experimentation platform where quantum phenomena such as the Meissner effect and flux pinning can be investigated.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty