Would you like to log in again?
Quantum Levitation Classroom Kit
Supplier: QUANTUM LEVITATION LTD
Ward's®
401
470306-210EA 3932.6 USD InStock
470306-210
Quantum Levitation Classroom Kit
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
The Classroom Kit will allows students to experiment with modern physics and learn about one of the most intriguing and exciting phenomenon of modern times – Superconductivity.
  • Easy to use
  • Highly durable
The kit contains a set of easy to conduct student experiments suitable for both high-school and undergraduate levels. The experiments and data analysis will enrich your student’s understanding and give them a taste of the work of true scientists.
Demonstrations and Student Activities:
Quantum Levitation.
The Meissner Effect.
Quantum Locking.
The Maglev Track.
Quantitative Experiments:
Measuring the Levitation Force.
Measuring the Critical Temperature Using Magnetic Levitation.
Superconductivity as a Mean to Investigate Classical Mechanics.
Ordering information: The Classroom Kit comes with Round maglev track Ø 40 cm (Ø15.7”)); Wooden stands; 3 handheld magnetic device; 1 medium Quantum Levitators; 1 small Quantum Levitators; 3 small Quantum Ed Levitators (thermometer included); Printed copy of “Quantum Levitation Booklet for Teachers“; and User manual.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty