Would you like to log in again?
Quantum Levitation Wave Flexible Track
Supplier: QUANTUM LEVITATION LTD
Ward's®
500-202 500-101
470306-220EA 2291.79 USD InStock
470306-220 470306-222
Quantum Levitation Wave Flexible Track
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Classical mechanics just became the most attractive experiment for students!
  • Easy to use
  • Highly durable
  • A teacher’s guide with more than 10 experiments in mechanics and modern physics is included with this kit
Unleash the powers of friction-less motion with Quantum Levitation with a flexible, adjustable magnetic track. The kit was specifically designed to support quantitative measurements of energy, velocity, acceleration and more.
Use video analysis software such as Vernier’s ‘Physics Video’ to analyse levitator motion above or below the track.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty