Would you like to log in again?
Garage Physics Ballista Kit, Eisco Scientific
Supplier: Eisco Scientific
GP00016
470307-348EA 132.5 USD InStock
470307-348
Garage Physics Ballista Kit, Eisco Scientific
Educational Materials Physics Educational Materials Mechanics Learning Activities
Uses torsion spring bundles and levers to launch ping pong balls.
  • No tools needed
  • Pre-cut pieces
  • Allows for multiple launch angles
Ballista Kit is assembled from pre-cut pieces with no tools. Uses torsion spring bundles and levers to launch a ping pong ball up to 30 feet. Allows for multiple launch angles and exploration of projectile motion and trajectories.
Delivery information: Includes 12 part kit, string, leather pouch, ping pong ball and guide.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty