Would you like to log in again?
Ward's® Manual Pipette Controller
Supplier: Ward's
Ward's®
P6420_WARD P6080-FI_WARD
470307-998PK 49.45 USD InStock
470307-998 470307-996
Ward's® Manual Pipette Controller
Pipettes Pipette Controllers
Ward's® Manual pipette controller is the precise and easy to use manual pipette controller that's always ready to use.
  • For glass or plastic pipettes
  • Accommodates pipettes up to 100 ml
  • Easy, single lever control
  • Autoclavable lower body
  • Fingertip speed control
After attaching a pipette, one squeeze of its large rubber filling bulb is all it takes to start pipetting. There are no wires, no tubing, and no battery to recharge.
Begin aspirating by touching the EZ-Flow™ lever which is designed to precisely control the filling speed, based on how gently or firmly the lever is pressed.
A built-in replaceable filter ensures optimum flow rate and protects the controller from liquid penetration and overfill. A separate button provides a positive blow-out function when required.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty