Would you like to log in again?
Chlorine Colorimeter, DC155-CL, LaMotte®
Supplier: LaMotte
3240 R-3240
470308-448EA 1033.6 USD InStock
470308-448 470308-446
Chlorine Colorimeter, DC155-CL, LaMotte®
Environmental Test Kits
The Lamotte Chlorine Colorimeter Kit, ideal for both field and lab use, covers the entire critical chlorine range of 0-4 ppm with an MDL of 0.03 ppm.
  • Chlorine Colorimeter Kit - DPD Tablet
  • Large graphical liquid crystal display
  • Simple, menu-driven operation
  • Lithium ion rechargeable battery and computer/wall adapter
  • IP67 Waterproof
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty