Would you like to log in again?
Ward's LiveSlides®
Supplier: VWR International
LS50BWB VWRI470313-232
470309-714BX 169.65 USD InStock
470309-714 470313-232
Ward's LiveSlides®
Slides Prepared Slides
Never before has it been so easy to view microorganisms, including bacteria, fungi, and protists.
  • Available as bulk pack of 50 slides only, or pack of 5 slides with transfer pipettes
  • Uniform depth of viewing well perfect for maintaining a shallow depth of field
  • Partitions help keep specimens in the viewing area
  • Polycarbonate material is safe for indoor or outdoor use
  • Can be used with traditional or digital microscopes
Each LiveSlide® consists of a base slide, and a set of coverslips of different thicknesses. One is very thin (0.13 mm), perfect for high magnification, and the second is a bit thicker (0.25 mm) for all applications. Each coverslip has a thin plastic film on each side for protection, one green and one clear. Remove both films by either peeling the film off or using a small piece of tape on the corner and lifting. Discard the films.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty